Assay Details
Gene Symbol: LOC100394788 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008991860.2 | XP_008990108.1 | 5 - 6 | 774 | 98 | |
XM_008991864.2 | XP_008990112.1 | 5 - 6 | 774 | 98 | ||
XM_008991868.2 | XP_008990116.1 | 5 - 6 | 795 | 98 | ||
XM_008991872.2 | XP_008990120.1 | 3 - 4 | 700 | 98 | ||
XM_008991877.2 | XP_008990125.1 | 4 - 5 | 1252 | 98 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
