Assay Details
| Gene Symbol: SELENOW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199908.2 | NP_001186837.1 | 4 - 5 | 320 | 63 | |
| NM_001278377.1 | NP_001265306.1 | 4 - 5 | 320 | 63 | ||
| GenBank mRNA | HX386774.1 | - | 4 - 5 | 267 | 63 | |
| HX387247.1 | - | 4 - 5 | 307 | 63 | ||
| HX387659.1 | - | 4 - 5 | 321 | 63 | ||
More Information
Set Membership: |
|

SELENOW
Genomic Map