Assay Details
Gene Symbol: CCDC155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008988390.2 | XP_008986638.1 | 7 - 8 | 844 | 60 | |
XM_017968184.1 | XP_017823673.1 | 7 - 8 | 844 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
