Assay Details
Gene Symbol: B3GNTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017972969.1 | XP_017828458.1 | 7 - 8 | 666 | 71 | |
XM_017972970.1 | XP_017828459.1 | 7 - 8 | 666 | 71 | ||
XM_017972971.1 | XP_017828460.1 | 7 - 8 | 666 | 71 | ||
XM_017972972.1 | XP_017828461.1 | 7 - 8 | 666 | 71 | ||
XM_017972973.1 | XP_017828462.1 | 8 - 9 | 2329 | 71 | ||
XM_017972974.1 | XP_017828463.1 | 5 - 6 | 501 | 71 | ||
XM_017972975.1 | XP_017828464.1 | 7 - 8 | 2229 | 71 | ||
XM_017972976.1 | XP_017828465.1 | 8 - 9 | 2316 | 71 | ||
XM_017972977.1 | XP_017828466.1 | 5 - 6 | 404 | 71 | ||
XM_017972978.1 | XP_017828467.1 | 4 - 5 | 1014 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
