Assay Details
| Gene Symbol: MFSD4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760715.4 | XP_002760761.1 | 3 - 4 | 777 | 66 | |
| XM_008985548.2 | XP_008983796.1 | 3 - 4 | 777 | 66 | ||
More Information
Set Membership: |
|

MFSD4A
Genomic Map