Assay Details
| Gene Symbol: LPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008979195.1 | XP_008977443.1 | 9 - 10 | 2073 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LPL| Gene Symbol: LPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008979195.1 | XP_008977443.1 | 9 - 10 | 2073 | 71 | |
Set Membership: |
|