Assay Details
Gene Symbol: NUBPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002753793.4 | XP_002753839.1 | 2 - 3 | 314 | 59 | |
XM_009005910.2 | XP_009004158.1 | 2 - 3 | 314 | 59 | ||
XM_009005911.2 | XP_009004159.1 | 2 - 3 | 314 | 59 | ||
XM_009005912.2 | XP_009004160.1 | 2 - 3 | 237 | 59 | ||
XM_009005913.1 | XP_009004161.1 | 2 - 3 | 232 | 59 | ||
XM_017977042.1 | XP_017832531.1 | 2 - 3 | 314 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
