Assay Details
Gene Symbol: CCDC106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002762517.3 | XP_002762563.1 | 3 - 4 | 636 | 72 | |
XM_008988684.2 | XP_008986932.1 | 4 - 5 | 1580 | 72 | ||
XM_008988685.2 | XP_008986933.1 | 4 - 5 | 1685 | 72 | ||
XM_008988686.2 | XP_008986934.1 | 4 - 5 | 1629 | 72 | ||
XM_017967306.1 | XP_017822795.1 | 5 - 6 | 1242 | 72 | ||
XM_017967307.1 | XP_017822796.1 | 4 - 5 | 1524 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
