Assay Details
| Gene Symbol: BRI3BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017976503.1 | XP_017831992.1 | 1 - 2 | 174 | 111 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017976503.1 transcripts.
Set Membership: |
|

BRI3BP
Genomic Map