Assay Details
Gene Symbol: CNNM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008980080.1 | XP_008978328.1 | 6 - 7 | 2204 | 57 | |
XM_017964071.1 | XP_017819560.1 | 7 - 8 | 2267 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
