Assay Details
| Gene Symbol: JHY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009007011.2 | XP_009005259.1 | 3 - 4 | 2648 | 121 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

JHY| Gene Symbol: JHY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009007011.2 | XP_009005259.1 | 3 - 4 | 2648 | 121 | |
Set Membership: |
|