Assay Details
Gene Symbol: MAPK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002744466.3 | XP_002744512.1 | 9 - 10 | 1318 | 68 | |
XM_008989579.2 | XP_008987827.1 | 9 - 10 | 1318 | 68 | ||
XM_008989587.2 | XP_008987835.1 | 9 - 10 | 1318 | 68 | ||
XM_008989592.2 | XP_008987840.1 | 9 - 10 | 1318 | 68 | ||
XM_017969426.1 | XP_017824915.1 | 10 - 11 | 1429 | 68 | ||
XM_017969427.1 | XP_017824916.1 | 10 - 11 | 1429 | 68 | ||
XM_017969428.1 | XP_017824917.1 | 10 - 11 | 1268 | 68 | ||
XM_017969430.1 | XP_017824919.1 | 11 - 12 | 1641 | 68 | ||
XM_017969432.1 | XP_017824921.1 | 10 - 11 | 1429 | 68 | ||
XM_017969434.1 | XP_017824923.1 | 10 - 11 | 1530 | 68 | ||
XM_017969436.1 | XP_017824925.1 | 9 - 10 | 1119 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
