Assay Details
Gene Symbol: NKIRAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002759656.4 | XP_002759702.1 | 3 - 4 | 630 | 102 | |
XM_017965895.1 | XP_017821384.1 | 2 - 3 | 268 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
