Assay Details
| Gene Symbol: CBR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761407.4 | XP_002761453.1 | 1 - 2 | 486 | 88 | |
| XM_008986643.2 | XP_008984891.1 | 1 - 2 | 486 | 88 | ||
More Information
Set Membership: |
|

CBR1
Genomic Map