Assay Details
Gene Symbol: LOC103791629 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017969948.1 | XP_017825437.1 | 4 | 1639 | 74 | |
XM_017969949.1 | XP_017825438.1 | 3 | 659 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
