Assay Details
| Gene Symbol: PRKAA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750871.3 | XP_002750917.1 | 6 - 7 | 861 | 72 | |
| XM_017974788.1 | XP_017830277.1 | 6 - 7 | 713 | 72 | ||
More Information
Set Membership: |
|

PRKAA2
Genomic Map