Assay Details
Gene Symbol: RPP40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002746253.3 | XP_002746299.1 | 7 - 8 | 1143 | 91 | |
XM_008994017.2 | XP_008992265.1 | 7 - 8 | 902 | 91 | ||
XM_017970936.1 | XP_017826425.1 | 6 - 7 | 1019 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
