Assay Details
| Gene Symbol: ZNF316 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017966877.1 | XP_017822366.1 | 2 - 3 | 514 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017966877.1 transcripts.
Set Membership: |
|

ZNF316
Genomic Map