Assay Details
| Gene Symbol: CIART | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759866.3 | XP_002759912.2 | 4 - 5 | 1362 | 64 | |
| XM_008984262.2 | XP_008982510.1 | 5 - 6 | 785 | 64 | ||
| XM_008984263.2 | XP_008982511.1 | 5 - 6 | 484 | 64 | ||
| XM_008984264.1 | XP_008982512.1 | 5 - 6 | 746 | 64 | ||
More Information
Set Membership: |
|

CIART
Genomic Map