Assay Details
| Gene Symbol: PDE6G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008997901.2 | XP_008996149.1 | 4 | 1258 | 102 | |
| XM_017972930.1 | XP_017828419.1 | 3 | 937 | 102 | ||
More Information
Set Membership: |
|

PDE6G
Genomic Map