Assay Details
Gene Symbol: IL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257233.1 | NP_001244162.1 | 2 - 3 | 179 | 80 | |
GenBank mRNA | DQ658152.1 | - | 2 - 3 | 179 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
