Assay Details
Gene Symbol: ZNF184 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002746112.3 | XP_002746158.1 | 5 - 6 | 816 | 71 | |
XM_008993801.2 | XP_008992049.1 | 5 - 6 | 645 | 71 | ||
XM_008993802.1 | XP_008992050.1 | 5 - 6 | 574 | 71 | ||
XM_017971579.1 | XP_017827068.1 | 5 - 6 | 897 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
