Assay Details
Gene Symbol: LOC100413262 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002744404.3 | XP_002744450.1 | 10 - 11 | 1300 | 102 | |
XM_008988341.2 | XP_008986589.1 | 10 - 11 | 1458 | 102 | ||
XM_008988348.2 | XP_008986596.1 | 11 - 12 | 1375 | 102 | ||
XM_008988357.2 | XP_008986605.1 | 9 - 10 | 1177 | 102 | ||
XM_017969030.1 | XP_017824519.1 | 9 - 10 | 1757 | 102 | ||
XM_017969036.1 | XP_017824525.1 | 7 - 8 | 1578 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
