Assay Details
Gene Symbol: LOC100413262 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002744404.3 | XP_002744450.1 | 2 - 3 | 291 | 74 | |
XM_008988341.2 | XP_008986589.1 | 2 - 3 | 449 | 74 | ||
XM_008988348.2 | XP_008986596.1 | 3 - 4 | 366 | 74 | ||
XM_008988357.2 | XP_008986605.1 | 1 - 2 | 168 | 74 | ||
XM_017969030.1 | XP_017824519.1 | 1 - 2 | 748 | 74 | ||
XM_017969035.1 | XP_017824524.1 | 1 - 2 | 749 | 74 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
