Assay Details
Gene Symbol: LOC100413262 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002744404.3 | XP_002744450.1 | 4 - 5 | 474 | 78 | |
XM_008988341.2 | XP_008986589.1 | 4 - 5 | 632 | 78 | ||
XM_008988348.2 | XP_008986596.1 | 5 - 6 | 549 | 78 | ||
XM_008988357.2 | XP_008986605.1 | 3 - 4 | 351 | 78 | ||
XM_017969030.1 | XP_017824519.1 | 3 - 4 | 931 | 78 | ||
XM_017969035.1 | XP_017824524.1 | 3 - 4 | 932 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
