Assay Details
Gene Symbol: LOC100413262 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002744404.3 | XP_002744450.1 | 6 - 7 | 649 | 75 | |
XM_008988341.2 | XP_008986589.1 | 6 - 7 | 807 | 75 | ||
XM_008988348.2 | XP_008986596.1 | 7 - 8 | 724 | 75 | ||
XM_008988357.2 | XP_008986605.1 | 5 - 6 | 526 | 75 | ||
XM_017969030.1 | XP_017824519.1 | 5 - 6 | 1106 | 75 | ||
XM_017969035.1 | XP_017824524.1 | 5 - 6 | 1107 | 75 | ||
XM_017969036.1 | XP_017824525.1 | 3 - 4 | 927 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
