Assay Details
Gene Symbol: UFSP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002745233.4 | XP_002745279.2 | 10 - 11 | 1329 | 64 | |
XM_008992385.2 | XP_008990633.1 | 10 - 11 | 1780 | 64 | ||
XM_017970105.1 | XP_017825594.1 | 10 - 11 | 1793 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
