Assay Details
Gene Symbol: BTN3A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017971580.1 | XP_017827069.1 | 1 - 2 | 126 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
