Assay Details
| Gene Symbol: SNX24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008991615.2 | XP_008989863.1 | 6 - 7 | 4509 | 67 | |
| XM_017969716.1 | XP_017825205.1 | 7 - 8 | 647 | 67 | ||
| XM_017969717.1 | XP_017825206.1 | 7 - 8 | 3040 | 67 | ||
| XM_017969718.1 | XP_017825207.1 | 7 - 8 | 1730 | 67 | ||
More Information
Set Membership: |
|

SNX24
Genomic Map