Assay Details
Gene Symbol: BHLHA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008996342.2 | XP_008994590.1 | 1 | 253 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
