Assay Details
Gene Symbol: C19H1orf131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002760899.4 | XP_002760945.2 | 3 - 4 | 568 | 101 | |
XM_008985806.2 | XP_008984054.1 | 3 - 4 | 571 | 101 |

More Information
Set Membership: |
![]() ![]() |
