Assay Details
Gene Symbol: HACD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002742969.4 | XP_002743015.1 | 3 - 4 | 360 | 89 | |
XM_008997638.2 | XP_008995886.1 | 3 - 4 | 359 | 89 | ||
XM_008997644.2 | XP_008995892.1 | 3 - 4 | 464 | 89 | ||
XM_017973884.1 | XP_017829373.1 | 3 - 4 | 359 | 89 | ||
XM_017973886.1 | XP_017829375.1 | 3 - 4 | 464 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
