Assay Details
Gene Symbol: APOPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009006602.2 | XP_009004850.1 | 1 - 2 | 224 | 113 | |
XM_017977363.1 | XP_017832852.1 | 1 - 2 | 224 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
