Assay Details
Gene Symbol: JAZF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002751446.2 | XP_002751492.1 | 3 - 4 | 627 | 107 | |
XM_009002227.2 | XP_009000475.1 | 3 - 4 | 556 | 107 |

More Information
Set Membership: |
![]() ![]() |
