Assay Details
Gene Symbol: HSD17B11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002745619.4 | XP_002745665.1 | 1 - 2 | 725 | 62 | |
XM_017970433.1 | XP_017825922.1 | 1 - 2 | 745 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
