Assay Details
Gene Symbol: TNFSF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205140.1 | NP_001192069.1 | 3 - 4 | 1105 | 61 | |
XM_008996551.2 | XP_008994799.1 | 3 - 4 | 1157 | 61 | ||
XM_008996552.2 | XP_008994800.1 | 3 - 4 | 1157 | 61 | ||
XM_008996553.2 | XP_008994801.1 | 3 - 4 | 1157 | 61 | ||
XM_008996554.2 | XP_008994802.1 | 3 - 4 | 1157 | 61 | ||
XM_008996555.2 | XP_008994803.1 | 3 - 4 | 1157 | 61 | ||
XM_008996556.2 | XP_008994804.1 | 3 - 4 | 1157 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
