Assay Details
Gene Symbol: TNFSF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205140.1 | NP_001192069.1 | 4 - 5 | 1228 | 74 | |
XM_008996551.2 | XP_008994799.1 | 4 - 5 | 1280 | 74 | ||
XM_008996552.2 | XP_008994800.1 | 4 - 5 | 1280 | 74 | ||
XM_008996553.2 | XP_008994801.1 | 4 - 5 | 1280 | 74 | ||
XM_008996554.2 | XP_008994802.1 | 4 - 5 | 1280 | 74 | ||
XM_008996555.2 | XP_008994803.1 | 4 - 5 | 1280 | 74 | ||
XM_008996556.2 | XP_008994804.1 | 4 - 5 | 1280 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
