Assay Details
| Gene Symbol: GATM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753420.4 | XP_002753466.1 | 1 - 2 | 385 | 56 | |
| XM_009005377.2 | XP_009003625.1 | 1 - 2 | 245 | 56 | ||
More Information
Set Membership: |
|

GATM
Genomic Map