Assay Details
| Gene Symbol: ECPAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009001202.2 | XP_008999450.1 | 47 - 48 | 5474 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ECPAS| Gene Symbol: ECPAS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009001202.2 | XP_008999450.1 | 47 - 48 | 5474 | 70 | |
Set Membership: |
|