Assay Details
Gene Symbol: KIAA1217 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009000038.2 | XP_008998286.2 | 16 - 17 | 3660 | 54 | |
XM_009000041.2 | XP_008998289.2 | 15 - 16 | 3577 | 54 | ||
XM_009000042.2 | XP_008998290.2 | 15 - 16 | 3552 | 54 | ||
XM_009000043.2 | XP_008998291.2 | 15 - 16 | 3575 | 54 | ||
XM_009000044.2 | XP_008998292.2 | 15 - 16 | 3551 | 54 | ||
XM_009000045.2 | XP_008998293.2 | 14 - 15 | 3467 | 54 | ||
XM_009000048.2 | XP_008998296.1 | 11 - 12 | 3473 | 54 | ||
XM_009000049.2 | XP_008998297.1 | 10 - 11 | 2631 | 54 | ||
XM_009000050.2 | XP_008998298.2 | 14 - 15 | 3499 | 54 | ||
XM_009000051.2 | XP_008998299.2 | 15 - 16 | 3623 | 54 | ||
XM_009000052.2 | XP_008998300.1 | 17 - 18 | 3869 | 54 | ||
XM_009000053.2 | XP_008998301.1 | 15 - 16 | 3625 | 54 | ||
XM_009000054.2 | XP_008998302.1 | 10 - 11 | 2581 | 54 | ||
XM_017974106.1 | XP_017829595.1 | 11 - 12 | 4449 | 54 | ||
XM_017974107.1 | XP_017829596.1 | 11 - 12 | 3473 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
