Assay Details
| Gene Symbol: UCP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754759.4 | XP_002754805.2 | 2 - 3 | 739 | 58 | |
| XM_009007452.2 | XP_009005700.1 | 2 - 3 | 745 | 58 | ||
More Information
Set Membership: |
|

UCP3
Genomic Map