Assay Details
Gene Symbol: CIDEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008982136.1 | XP_008980384.1 | 2 - 3 | 147 | 64 | |
XM_008982137.1 | XP_008980385.1 | 2 - 3 | 103 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
