Assay Details
Gene Symbol: SIL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008985639.2 | XP_008983887.1 | 7 - 8 | 961 | 67 | |
XM_008985651.2 | XP_008983899.1 | 7 - 8 | 795 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
