Assay Details
| Gene Symbol: APOBEC3H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017963597.1 | XP_017819086.1 | 3 - 4 | 519 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

APOBEC3H| Gene Symbol: APOBEC3H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017963597.1 | XP_017819086.1 | 3 - 4 | 519 | 94 | |
Set Membership: |
|