Assay Details
| Gene Symbol: LOC100409586 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746172.3 | XP_002746218.2 | 1 - 2 | 360 | 75 | |
| XM_008993844.2 | XP_008992092.1 | 1 - 2 | 103 | 75 | ||
| XM_008993845.2 | XP_008992093.1 | 1 - 2 | 457 | 75 | ||
| XM_008993846.2 | XP_008992094.1 | 1 - 2 | 457 | 75 | ||
| XM_008993847.2 | XP_008992095.1 | 1 - 2 | 457 | 75 | ||
| XM_008993848.2 | XP_008992096.1 | 1 - 2 | 457 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008993844.2 transcripts.
Set Membership: |
|

LOC100409586
Genomic Map