Assay Details
Gene Symbol: CDCA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002749330.3 | XP_002749376.1 | 8 - 9 | 1316 | 70 | |
XM_003733164.3 | XP_003733212.1 | 7 - 8 | 1091 | 70 | ||
XM_017973404.1 | XP_017828893.1 | 7 - 8 | 1233 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
