Assay Details
Gene Symbol: TMEM9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002755019.4 | XP_002755065.1 | 4 - 5 | 1211 | 78 | |
XM_017977891.1 | XP_017833380.1 | 3 - 4 | 1119 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
