Assay Details
| Gene Symbol: POLR3G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008991807.2 | XP_008990055.1 | 6 - 7 | 648 | 108 | |
| XM_008991808.2 | XP_008990056.1 | 6 - 7 | 644 | 108 | ||
More Information
Set Membership: |
|

POLR3G
Genomic Map