Assay Details
Gene Symbol: RALYL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003734963.3 | XP_003735011.1 | 6 - 7 | 1651 | 94 | |
XM_003734964.3 | XP_003735012.1 | 6 - 7 | 1121 | 94 | ||
XM_003734965.2 | XP_003735013.1 | 5 - 6 | 474 | 94 | ||
XM_003734966.3 | XP_003735014.1 | 5 - 6 | 1618 | 94 | ||
XM_008982843.2 | XP_008981091.1 | 6 - 7 | 1127 | 94 | ||
XM_008982844.2 | XP_008981092.1 | 5 - 6 | 1088 | 94 | ||
XM_008982845.1 | XP_008981093.1 | 6 - 7 | 969 | 94 | ||
XM_008982846.2 | XP_008981094.1 | 6 - 7 | 869 | 94 | ||
XM_008982848.2 | XP_008981096.1 | 6 - 7 | 1134 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
