Assay Details
Gene Symbol: CNOT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002749786.3 | XP_002749832.1 | 2 - 3 | 583 | 75 | |
XM_008999491.2 | XP_008997739.1 | 2 - 3 | 584 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
